missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DRP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26631-1-AP-150UL
This item is not returnable.
View return policy
Description
Members of the dystrophin family of proteins perform a critical role in the maintece of membrane-associated complexes at points of intercellular contact in vertebrate cells. Dystrophin related protein 2 is predicted to resemble certain short C-terminal isoforms of dystrophin and dystrophin-related protein 1 (DRP1 or utrophin). DRP2 is expressed principally in the brain and spinal cord.Specifications
| DRP2 | |
| Polyclonal | |
| Unconjugated | |
| DRP2 | |
| DRP 2, DRP2, dystrophin related protein 2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 13497, 1821, 66027 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.4 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q05AA6, Q13474, Q9EPA0 | |
| DRP2 | |
| DRP2 Fusion Protein Ag24462 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction