Learn More
Dextromethorphan hydrobromide, Thermo Scientific™
£75.00 - £579.00
Chemical Identifiers
| CAS | 125-69-9 |
|---|---|
| Molecular Formula | C18H26BrNO |
| Molecular Weight (g/mol) | 352.316 |
| MDL Number | MFCD02173901 |
| InChI Key | MISZALMBODQYFT-URVXVIKDSA-N |
| Synonym | dextromethorphan hydrobromide, antussan, demorphan, dormethan, metrorat, medicon, tusilan, tussade, d-methorphan hydrobromide, romilar |
| PubChem CID | 5464025 |
| SMILES | CN1CCC23CCCCC2C1CC4=C3C=C(C=C4)OC.Br |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15474269
|
Thermo Scientific Alfa Aesar
J61732.06 |
5g |
£75.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15484269
|
Thermo Scientific Alfa Aesar
J61732.09 |
10g |
£154.00
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15494269
|
Thermo Scientific Alfa Aesar
J61732.18 |
50g |
£579.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsIt is used as a potentially useful NMDA receptor agonist. it is usually used as central nervous antitussive, cough suppressant. This is one of the most popular ingredients in over-the-counter cold and cough medications. Dextromethorphan hydrobromide may be added to other preparations to help treat pain and a variety of other disturbances as well. It is used as a pharmaceutical raw material.
Solubility
Soluble in water (40 mM), and alcohol (1:10).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Chemical Identifiers
| C18H26BrNO | |
| MFCD02173901 | |
| dextromethorphan hydrobromide, antussan, demorphan, dormethan, metrorat, medicon, tusilan, tussade, d-methorphan hydrobromide, romilar | |
| CN1CCC23CCCCC2C1CC4=C3C=C(C=C4)OC.Br |
Specifications
| 125-69-9 | |
| MFCD02173901 | |
| 14,8091 | |
| Soluble in water (40 mM),and alcohol (1:10). | |
| CN1CCC23CCCCC2C1CC4=C3C=C(C=C4)OC.Br | |
| 5464025 | |
| Dextromethorphan hydrobromide |
| C18H26BrNO | |
| 6453793 | |
| dextromethorphan hydrobromide, antussan, demorphan, dormethan, metrorat, medicon, tusilan, tussade, d-methorphan hydrobromide, romilar | |
| MISZALMBODQYFT-URVXVIKDSA-N | |
| 352.316 | |
| 352.32 |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
missing translation for 'einecsNumber' : 204-750-1
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
- Dextromethorphan hydrobromide
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
Hazard Statement
- H302-Harmful if swallowed.
Precautionary Statement
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
- MIXTURE LIST-Contain: Dextromethorphan hydrobromide
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.