missing translation for 'onlineSavingsMsg'
Learn More

Dextromethorphan hydrobromide

Product Code. 15494269
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
Unit Size:
10g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15494269 50 g 50g
15474269 5 g 5g
15484269 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15494269 Supplier Thermo Scientific Alfa Aesar Supplier No. J61732.18
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 125-69-9 | C18H26BrNO | 352.316 g/mol

It is used as a potentially useful NMDA receptor agonist. it is usually used as central nervous antitussive, cough suppressant. This is one of the most popular ingredients in over-the-counter cold and cough medications. Dextromethorphan hydrobromide may be added to other preparations to help treat pain and a variety of other disturbances as well. It is used as a pharmaceutical raw material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a potentially useful NMDA receptor agonist. it is usually used as central nervous antitussive, cough suppressant. This is one of the most popular ingredients in over-the-counter cold and cough medications. Dextromethorphan hydrobromide may be added to other preparations to help treat pain and a variety of other disturbances as well. It is used as a pharmaceutical raw material.

Solubility
Soluble in water (40 mM), and alcohol (1:10).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 125-69-9
Molecular Formula C18H26BrNO
Molecular Weight (g/mol) 352.316
MDL Number MFCD02173901
InChI Key MISZALMBODQYFT-URVXVIKDSA-N
Synonym dextromethorphan hydrobromide, antussan, demorphan, dormethan, metrorat, medicon, tusilan, tussade, d-methorphan hydrobromide, romilar
PubChem CID 5464025
SMILES CN1CCC23CCCCC2C1CC4=C3C=C(C=C4)OC.Br

Specifications

Melting Point 125°C to 127°C
Color White
Quantity 50 g
Beilstein 6453793
Merck Index 14,8091
Solubility Information Soluble in water (40 mM),and alcohol (1:10).
Formula Weight 352.32
Physical Form Solid
Chemical Name or Material Dextromethorphan hydrobromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.