missing translation for 'onlineSavingsMsg'
Learn More

Dextromethorphan hydrobromide, Thermo Scientific™

Brand:  Thermo Scientific Alfa Aesar J61732.09

Quantity 10g

 View more versions of this product

Product Code. 15484269

  • £154.00 / 10g

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used as a potentially useful NMDA receptor agonist. it is usually used as central nervous antitussive, cough suppressant. This is one of the most popular ingredients in over-the-counter cold and cough medications. Dextromethorphan hydrobromide may be added to other preparations to help treat pain and a variety of other disturbances as well. It is used as a pharmaceutical raw material.

Solubility
Soluble in water (40 mM), and alcohol (1:10).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

125-69-9
352.316
MISZALMBODQYFT-URVXVIKDSA-N
5464025
C18H26BrNO
MFCD02173901
dextromethorphan hydrobromide, antussan, demorphan, dormethan, metrorat, medicon, tusilan, tussade, d-methorphan hydrobromide, romilar
CN1CCC23CCCCC2C1CC4=C3C=C(C=C4)OC.Br

Specifications

125-69-9
MFCD02173901
6453793
dextromethorphan hydrobromide, antussan, demorphan, dormethan, metrorat, medicon, tusilan, tussade, d-methorphan hydrobromide, romilar
MISZALMBODQYFT-URVXVIKDSA-N
352.316
352.32
C18H26BrNO
10g
14,8091
Soluble in water (40 mM),and alcohol (1:10).
CN1CCC23CCCCC2C1CC4=C3C=C(C=C4)OC.Br
5464025
Dextromethorphan hydrobromide
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.