missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Desmoglein-3 Mouse, Clone: DSG3/1535, enQuireBio™
Mouse Monoclonal Antibody
£181.83 - £395.00
Specifications
Antigen | Desmoglein-3 |
---|---|
Clone | DSG3/1535 |
Concentration | 200μg/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15969907
|
EnquireBio
1830-MSM5-P1 |
100 μg |
£395.00
100µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15959907
|
EnquireBio
1830-MSM5-P0 |
20 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
Desmoglein-3 | |
200μg/mL | |
Monoclonal | |
Mouse | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
1830 | |
Recombinant human DSG3 extracellular protein fragment | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
130kDa |
DSG3/1535 | |
Immunohistochemistry | |
Unconjugated | |
RUO | |
NP_001935 | |
DSG3 | |
IgG1 κ | |
Protein A/G Purified | |
Cell surface | |
A431 cells. Skin or Lung squamous cell carcinoma. |