missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Desmoglein-3 Mouse, Clone: DSG3/1535, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 1830-MSM5-P0
Additional Details : Weight : 0.01000kg
Specifications
Desmoglein-3 | |
Cell surface | |
DSG3/1535 | |
Unconjugated | |
NP_001935 | |
Mouse | |
130kDa | |
Protein A/G Purified | |
RUO | |
1830 | |
IgG1 κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
DSG3 | |
Recombinant human DSG3 extracellular protein fragment | |
A431 cells. Skin or Lung squamous cell carcinoma. | |
20 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |