missing translation for 'onlineSavingsMsg'
Learn More

DBPA Antibody (PCRP-YBX3-2D12), Novus Biologicals™

Mouse Monoclonal Antibody

£187.00 - £351.00

Specifications

Antigen DBPA
Clone PCRP-YBX3-2D12
Concentration 0.2 mg/mL
Dilution Western Blot : 1-2 μg/mL, Flow Cytometry : 1-2 μg/million cells, Immunocytochemistry/ Immunofluorescence : 1-2 μg/mL, Immunoprecipitation : 1-2ug/100-500ug protein, Immunohistochemistry-Paraffin : 1-2 μg/mL
Applications Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18663164
View Documents
Novus Biologicals
NBP3-23901-20ug
20 μg
£187.00
20µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18673765
View Documents
Novus Biologicals
NBP3-23901-100ug
100 μg
£351.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

DBPA Monoclonal antibody specifically detects DBPA in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications

Specifications

DBPA
0.2 mg/mL
Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Human
cold shock domain protein A, Cold shock domain-containing protein A, cold-shock domain protein A, CSDA1, dbpA, DBPAcold-shock domain containing A1, DNA-binding protein A, Single-strand DNA-binding protein NF-GMB, ZO-1-associated nucleic acid-binding protein, ZONAB
Recombinant full-length human DBPA protein
Primary
Store at 4°C.
PCRP-YBX3-2D12
Western Blot : 1-2 μg/mL, Flow Cytometry : 1-2 μg/million cells, Immunocytochemistry/ Immunofluorescence : 1-2 μg/mL, Immunoprecipitation : 1-2ug/100-500ug protein, Immunohistochemistry-Paraffin : 1-2 μg/mL
Monoclonal
Purified
RUO
10 mM PBS with 0.05% BSA
8531
IgG2b
Protein A or G purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.