missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DBPA Antibody (PCRP-YBX3-2D12), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-23901-100ug
This item is not returnable.
View return policy
Description
DBPA Monoclonal antibody specifically detects DBPA in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications
| DBPA | |
| Monoclonal | |
| 0.2 mg/mL | |
| Western Blot : 1-2 μg/mL, Flow Cytometry : 1-2 μg/million cells, Immunocytochemistry/ Immunofluorescence : 1-2 μg/mL, Immunoprecipitation : 1-2ug/100-500ug protein, Immunohistochemistry-Paraffin : 1-2 μg/mL | |
| cold shock domain protein A, Cold shock domain-containing protein A, cold-shock domain protein A, CSDA1, dbpA, DBPAcold-shock domain containing A1, DNA-binding protein A, Single-strand DNA-binding protein NF-GMB, ZO-1-associated nucleic acid-binding protein, ZONAB | |
| Recombinant full-length human DBPA protein | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| PCRP-YBX3-2D12 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 8531 | |
| Store at 4°C. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction