missing translation for 'onlineSavingsMsg'
Learn More

DBPA Antibody (PCRP-YBX3-2D12), Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP3-23901-20ug

 View more versions of this product

Product Code. 18663164

  • £187.00 / 20µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

DBPA Monoclonal antibody specifically detects DBPA in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

DBPA
Monoclonal
0.2 mg/mL
Western Blot : 1-2 μg/mL, Flow Cytometry : 1-2 μg/million cells, Immunocytochemistry/ Immunofluorescence : 1-2 μg/mL, Immunoprecipitation : 1-2ug/100-500ug protein, Immunohistochemistry-Paraffin : 1-2 μg/mL
cold shock domain protein A, Cold shock domain-containing protein A, cold-shock domain protein A, CSDA1, dbpA, DBPAcold-shock domain containing A1, DNA-binding protein A, Single-strand DNA-binding protein NF-GMB, ZO-1-associated nucleic acid-binding protein, ZONAB
Recombinant full-length human DBPA protein
20 μg
Primary
Human
Purified
Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
PCRP-YBX3-2D12
Unconjugated
10 mM PBS with 0.05% BSA
Mouse
Protein A or G purified
RUO
8531
Store at 4°C.
IgG2b
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.