missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYP26A1 Mouse anti-Human, Clone: F27 P6 A1, Novus Biologicals™
Mouse Monoclonal Antibody
£198.00 - £401.00
Specifications
| Antigen | CYP26A1 |
|---|---|
| Clone | F27 P6 A1 |
| Dilution | Western Blot 1 - 2 ug/ml, ELISA, Immunohistochemistry |
| Applications | Western Blot, ELISA, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18322572
|
Bio-Techne
NBP3-11915-25UG |
25 μg |
£198.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18315452
|
Novus Biologicals
NBP3-11915-200UG |
200 μg |
£401.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CYP26A1 Monoclonal antibody specifically detects CYP26A1 in Human samples. It is validated for Western Blot, ELISA, ImmunohistochemistrySpecifications
| CYP26A1 | |
| Western Blot 1 - 2 ug/ml, ELISA, Immunohistochemistry | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CP26, CYP26Retinoic acid 4-hydroxylase, cytochrome P450 26A1, Cytochrome P450 retinoic acid-inactivating 1, cytochrome P450, family 26, subfamily A, polypeptide 1, cytochrome P450, subfamily XXVIA, polypeptide 1, Cytochrome P450RAI, EC 1.14, hP450RAI, P450, retinoic acid-inactivating, 1, P450RAI, P450RAI1Retinoic acid-metabolizing cytochrome | |
| Hybridoma produced by the fusion of splenocytes from BALB/c mice immunized with a synthetic peptide derived from the C-terminus of the human CYP450 26A1 protein and mouse myeloma Ag8563 cells. (Uniprot: O43174) | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
| F27 P6 A1 | |
| Western Blot, ELISA, Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Stem Cell Markers | |
| PBS | |
| 1592 | |
| IgG2b | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title