missing translation for 'onlineSavingsMsg'
Learn More

CYP26A1 Mouse anti-Human, Clone: F27 P6 A1, Novus Biologicals™

Product Code. 18322572 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
200 μg
25 μg
Unit Size:
200µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18322572 25 μg 25µg
18315452 200 μg 200µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18322572 Supplier Bio-Techne Supplier No. NBP31191525UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CYP26A1 Monoclonal antibody specifically detects CYP26A1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYP26A1
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone F27 P6 A1
Conjugate Unconjugated
Dilution Western Blot 1 - 2 ug/ml, ELISA, Immunohistochemistry
Formulation PBS
Gene Alias CP26, CYP26Retinoic acid 4-hydroxylase, cytochrome P450 26A1, Cytochrome P450 retinoic acid-inactivating 1, cytochrome P450, family 26, subfamily A, polypeptide 1, cytochrome P450, subfamily XXVIA, polypeptide 1, Cytochrome P450RAI, EC 1.14, hP450RAI, P450, retinoic acid-inactivating, 1, P450RAI, P450RAI1Retinoic acid-metabolizing cytochrome
Host Species Mouse
Immunogen Hybridoma produced by the fusion of splenocytes from BALB/c mice immunized with a synthetic peptide derived from the C-terminus of the human CYP450 26A1 protein and mouse myeloma Ag8563 cells. (Uniprot: O43174)
Purification Method Protein A or G purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 1592
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.