missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYP26A1 Mouse anti-Human, Clone: F27 P6 A1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-11915-200UG
This item is not returnable.
View return policy
Description
CYP26A1 Monoclonal antibody specifically detects CYP26A1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry
Specifications
| CYP26A1 | |
| Monoclonal | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry | |
| F27 P6 A1 | |
| Western Blot 1 - 2 ug/ml, ELISA, Immunohistochemistry | |
| CP26, CYP26Retinoic acid 4-hydroxylase, cytochrome P450 26A1, Cytochrome P450 retinoic acid-inactivating 1, cytochrome P450, family 26, subfamily A, polypeptide 1, cytochrome P450, subfamily XXVIA, polypeptide 1, Cytochrome P450RAI, EC 1.14, hP450RAI, P450, retinoic acid-inactivating, 1, P450RAI, P450RAI1Retinoic acid-metabolizing cytochrome | |
| Hybridoma produced by the fusion of splenocytes from BALB/c mice immunized with a synthetic peptide derived from the C-terminus of the human CYP450 26A1 protein and mouse myeloma Ag8563 cells. (Uniprot: O43174) | |
| 200 μg | |
| Stem Cell Markers | |
| 1592 | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction