missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Cyclin E1 Antibody (168), Novus Biologicals™
Mouse Monoclonal Antibody
£162.00 - £386.00
Specifications
| Antigen | Cyclin E1 |
|---|---|
| Clone | 168 |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30066908
|
Novus Biologicals™
NBP3-30974-25ug |
25 μg |
£162.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30066843
|
Novus Biologicals™
NBP3-30974-100ug |
100 μg |
£386.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cyclin E1 Monoclonal antibody specifically detects Cyclin E1 in Human samples. It is validated for Immunoprecipitation,Western BlotSpecifications
| Cyclin E1 | |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Mouse | |
| Cancer, Cell Cycle and Replication, Cellular Markers, Ovarian Carcinoma Cell Markers, Phospho Specific | |
| Phosphate buffered saline | |
| 898 | |
| IgG2a | |
| Protein A or G purified |
| 168 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CCNEcyclin Es, cyclin E1, cyclin Et, G1/S-specific cyclin-E1 | |
| Recombinant full-length human Cyclin E1 protein | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title