missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Cyclin E1 Antibody (168), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-30974-25ug
This item is not returnable.
View return policy
Description
Cyclin E1 Monoclonal antibody specifically detects Cyclin E1 in Human samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| Cyclin E1 | |
| Monoclonal | |
| Unconjugated | |
| CCNEcyclin Es, cyclin E1, cyclin Et, G1/S-specific cyclin-E1 | |
| Recombinant full-length human Cyclin E1 protein | |
| 25 μg | |
| Cancer, Cell Cycle and Replication, Cellular Markers, Ovarian Carcinoma Cell Markers, Phospho Specific | |
| 898 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG2a |
| Immunoprecipitation, Western Blot | |
| 168 | |
| Phosphate buffered saline | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction