missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Cyclin E1 Antibody (168), Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals™ NBP3-30974-100ug

 View more versions of this product

Product Code. 30066843

  • £386.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Cyclin E1 Monoclonal antibody specifically detects Cyclin E1 in Human samples. It is validated for Immunoprecipitation,Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cyclin E1
Monoclonal
Unconjugated
CCNEcyclin Es, cyclin E1, cyclin Et, G1/S-specific cyclin-E1
Recombinant full-length human Cyclin E1 protein
100 μg
Cancer, Cell Cycle and Replication, Cellular Markers, Ovarian Carcinoma Cell Markers, Phospho Specific
898
Store at -20°C. Avoid freeze/thaw cycles.
IgG2a
Immunoprecipitation, Western Blot
168
Phosphate buffered saline
Mouse
Protein A or G purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.