missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRK Mouse, anti-Human, Biotin, Clone: 2B7E1, Proteintech
Mouse Monoclonal Antibody
£135.00 - £375.00
Specifications
| Antigen | CRK |
|---|---|
| Clone | 2B7E1 |
| Concentration | 1000 μg/mL |
| Applications | ELISA |
| Conjugate | Biotin |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18006875
|
Proteintech
BIOTIN-67035-100UL |
100 μL |
£375.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18090576
|
Proteintech
BIOTIN-67035-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The CRK antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CRK. This antibody recognizes human, mouse, rat antigen. The CRK antibody has been validated for the following applications: ELISA analysis. This antibody is biotin.
Specifications
| CRK | |
| 1000 μg/mL | |
| Biotin | |
| Mouse | |
| 1398 | |
| IgG1 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
| 2B7E1 | |
| ELISA | |
| Liquid | |
| Human, Mouse, Rat | |
| Adapter molecule crk, CRK, CRKII, p38, Proto oncogene c Crk | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title