missing translation for 'onlineSavingsMsg'
Learn More

CRK Mouse, anti-Human, Biotin, Clone: 2B7E1, Proteintech

Product Code. 18006875 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
100 μL
20 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18006875 100 μL 100µL
18090576 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18006875 Supplier Proteintech Supplier No. BIOTIN67035100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The CRK antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CRK. This antibody recognizes human, mouse, rat antigen. The CRK antibody has been validated for the following applications: ELISA analysis. This antibody is biotin.

Specifications

Antigen CRK
Applications ELISA
Clone 2B7E1
Concentration 1000 μg/mL
Conjugate Biotin
Gene Symbols Adapter molecule crk, CRK, CRKII, p38, Proto oncogene c Crk
Host Species Mouse
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 1398
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.