Learn More
CRK Mouse, anti-Human, Biotin, Clone: 2B7E1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech BIOTIN-67035-20UL
Description
The CRK antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CRK. This antibody recognizes human, mouse, rat antigen. The CRK antibody has been validated for the following applications: ELISA analysis. This antibody is biotin.
Specifications
| CRK | |
| 2B7E1 | |
| Biotin | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
| ELISA | |
| 1000 μg/mL | |
| Adapter molecule crk, CRK, CRKII, p38, Proto oncogene c Crk | |
| 20 μL | |
| 1398 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| IgG1 |