missing translation for 'onlineSavingsMsg'
Learn More

Corning™ PureCoat™ Carboxyl Rectangular Canted Neck Cell Culture Flasks with Vented Cap

Negatively charged surface is ideal for for preparative cell culture, cell-based, and HTS screening assays

Brand:  Corning™ 354778

Code : 66

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 13456839

  • £36.72 / Pack of 5
Estimated Shipment: 06-09-2024
to see stock.
Add to basket

Description

Description

Corning™ PureCoat™ amine (positively charged) and carboxyl (negatively charged) surfaces provide improved cell attachment, faster cell proliferation, and enhanced recovery post-thaw over standard TC surfaces. These surfaces function with a broad range of primary, transfected, transformed, and fastidious cell types, and have demonstrated utility in serum-reduced or serum-free conditions.
Specifications

Specifications

8.45 oz.
Polystyrene
Canted
5 Pk.
Carboxyl
75 cm2
250 mL
For preparative cell culture, cell-based and HTS screening assays
5/Cs.
Rectangular
Vented
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Disclaimer: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.