missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ PureCoat™ Carboxyl Rectangular Canted Neck Cell Culture Flasks with Vented Cap



missing translation for 'orderingAttributeHoverText'
Capacity (Metric):
250 mL
750 mL
No. per Case:
5/Cs.
50/Cs.
missing translation for 'unitSize'
Pack of 5
Pack of 50
Description
Corning™ PureCoat™ amine (positively charged) and carboxyl (negatively charged) surfaces provide improved cell attachment, faster cell proliferation, and enhanced recovery post-thaw over standard TC surfaces. These surfaces function with a broad range of primary, transfected, transformed, and fastidious cell types, and have demonstrated utility in serum-reduced or serum-free conditions.
Specifications
Specifications
| Capacity (English) | 8.45 oz. |
| Capacity (Metric) | 250 mL |
| Material | Polystyrene |
| For Use With (Application) | For preparative cell culture, cell-based and HTS screening assays |
| Neck Style | Canted |
| No. per Case | 50/Cs. |
| No. per Pack | 5 Pk. |
| Shape | Rectangular |
| Surface Coating | Carboxyl |
| Closure Type | Vented |
| Show More |
Disclaimer: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction