Learn More
Macherey-Nagel™ CHROMABOND™ Multi 96 PP Reservoir Collection Vessels, 12 x 8-well Strips
Product Code. 10610162 Shop All Macherey-Nagel Products
Change view
Click to view available options
Quantity:
10 Pk.
Unit Size:
Pack of 10
Product Code. Quantity Unit Size
10610162 10 Pk. Pack of 10
1 options
Product Code. 10610162 Supplier Macherey-Nagel™ Supplier No. 738652
Estimated Shipment: 22-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The Reservoir Collection Vessels are used as refills for the CHROMABOND™ Multi 96 Collection Rack for the Multi 96 Vacuum Manifold in solid phase extraction (SPE).
  • Accessory 12 x 8-well strip refills
  • For Multi 96 collection rack
  • Polypropylene
Specifications
Includes 12 x 8-well Roller Strips
Capacity (Metric) 1.0 mL (each tube)
For Use With (Application) Solid phase extraction vacuum manifold Multi 96
Material Polypropylene
Quantity 10 Pk.
Product Type Collecting Vessel