missing translation for 'onlineSavingsMsg'
Learn More

Collagen II Ab-2 Mouse Monoclonal Antibody, Epredia™

Ensure accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence and flow cytometry experiments.

£120.36 - £703.80

Specifications

Antigen Collagen II Ab-2
Clone 2B1.5
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Host Species Mouse
View More Specs

Products 9
Product Code Brand Form Conjugate Quantity Price Quantity & Availability  
Product Code Brand Form Conjugate Quantity Price Quantity & Availability  
12620657
View Documents Promotion Details
Epredia
MS-235-B0
Concentrated Biotin 100 μL
£181.00
100µL
View alternative products
Save Up to 
»
Estimated Shipment: 10-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
12630657
View Documents Promotion Details
Epredia
MS-235-B1
Concentrated Biotin 500 μL
£507.00
500µL
View alternative products
Save Up to 
»
Estimated Shipment: 10-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
12660657
View Documents Promotion Details
Epredia
MS-235-P1
Concentrated Unconjugated 500 μL
£393.00
500µL
View alternative products
Save Up to 
»
Estimated Shipment: 10-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
12640657
View Documents Promotion Details
Epredia
MS-235-P
Concentrated Unconjugated 1 mL
£626.00
1mL
View alternative products
Save Up to 
»
Estimated Shipment: 10-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
12670657
View Documents Promotion Details
Epredia
MS-235-P1ABX
Concentrated Unconjugated 100 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12680657
View Documents Promotion Details
Epredia
MS-235-PABX
Concentrated Unconjugated 200 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12610657
View Documents Promotion Details
Epredia
MS-235-B
Concentrated Biotin 1 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12650657
View Documents Promotion Details
Epredia
MS-235-P0
Concentrated Unconjugated 100 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12610667
View Documents Promotion Details
Epredia
MS-235-R7
Prediluted Unconjugated 7 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Host Species: Mouse

Clone: 2B1.5

Isotype: IgG2a/κ

Species Reactivity: Human, Mouse, Rat, Cow and Chicken. Others not known.

Epitope: Carboxyl-terminal one quarter of the type II collagen. The epitope for 2B1.5 MAb is not susceptible to digestion of lathyritic type II collagen with recombinant stromelysin

Immunogen: A purified preparation of lathyritic type II collagen from embryonic chicken sternum

Positive Control: Cartilage in lung or fetus

Cellular Localization: Cartilage matrix

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Collagen II Ab-2
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Mouse
Extracellular Matrix
A purified preparation of lathyritic type II collagen from embryonic chicken sternum
Primary
2B1.5
Monoclonal
RUO
Bovine, Chicken, Human, Mouse, Rat
IgG2a κ
Videos
SDS
Documents

Documents

Product Certifications