Learn More
Collagen II Ab-2 Mouse Monoclonal Antibody, Epredia™
Ensure accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence and flow cytometry experiments.
Brand: Epredia MS-235-B1
Code : LD
Additional Details : Weight : 0.00190kg
Description
Host Species: Mouse
Clone: 2B1.5
Isotype: IgG2a/κ
Species Reactivity: Human, Mouse, Rat, Cow and Chicken. Others not known.
Epitope: Carboxyl-terminal one quarter of the type II collagen. The epitope for 2B1.5 MAb is not susceptible to digestion of lathyritic type II collagen with recombinant stromelysin
Immunogen: A purified preparation of lathyritic type II collagen from embryonic chicken sternum
Positive Control: Cartilage in lung or fetus
Cellular Localization: Cartilage matrix
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
- Flow Cytometry
Note:
USA: RUO; Int'l: RUO
Specifications
Collagen II Ab-2 | |
Monoclonal | |
200μg/mL | |
Mouse | |
500 μL | |
Extracellular Matrix | |
Bovine, Chicken, Human, Mouse, Rat | |
IgG2a κ |
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
2B1.5 | |
Biotin | |
A purified preparation of lathyritic type II collagen from embryonic chicken sternum | |
RUO | |
Primary | |
Concentrated |