missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cofilin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£150.00 - £389.00
Specifications
| Antigen | Cofilin |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin 1:50-1:100 |
| Applications | Immunohistochemistry (Paraffin), Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18235802
|
Novus Biologicals
NB100-81866-0.025ML |
0.025 mL |
£150.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18756193
|
Novus Biologicals
NB100-81866 |
0.1 mL |
£389.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cofilin Polyclonal specifically detects Cofilin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Cofilin | |
| Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin 1:50-1:100 | |
| Polyclonal | |
| Rabbit | |
| Cytoskeleton Markers | |
| P23528 | |
| 1072 | |
| Peptide sequence around aa. 1∼5 (M-A-S-G-V) derived from Human cofilin. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 1.0 mg/mL | |
| Immunohistochemistry (Paraffin), Immunohistochemistry, Western Blot | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat | |
| 18 kDa phosphoprotein, CFL, cofilin 1 (non-muscle), Cofilin, non-muscle isoform, cofilin-1, p18 | |
| CFL1 | |
| IgG | |
| Affinity Purified | |
| 19 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title