Learn More
Cofilin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-81866
This item is not returnable.
View return policy
Description
Cofilin Polyclonal specifically detects Cofilin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Rabbit | |
| Peptide sequence around aa. 1∼5 (M-A-S-G-V) derived from Human cofilin. | |
| 0.1 mL |