missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cofilin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-81866-0.025ML
This item is not returnable.
View return policy
Description
Cofilin Polyclonal specifically detects Cofilin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Cofilin | |
| Polyclonal | |
| Unconjugated | |
| phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, and 50% glycerol. with 0.02% Sodium Azide | |
| 18 kDa phosphoprotein, CFL, cofilin 1 (non-muscle), Cofilin, non-muscle isoform, cofilin-1, p18 | |
| Rabbit | |
| 19 kDa | |
| 0.025 mL | |
| Cytoskeleton Markers | |
| 1072 | |
| Human, Mouse, Rat | |
| IgG |
| Immunohistochemistry (Paraffin), Immunohistochemistry, Western Blot | |
| 1.0 mg/mL | |
| Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin 1:50-1:100 | |
| P23528 | |
| CFL1 | |
| Peptide sequence around aa. 1∼5 (M-A-S-G-V) derived from Human cofilin. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| cofilin (specific to a.a. 1022) detects endogenous levels of total cofilin protein. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction