Learn More
Zwiebel™ Class M1 Box Weights with Certificate
Use with weighing instruments of medium accuracy class III. Zwiebel™ Class M1 Box Weights are ideal for the verification or calibration of class M₂ weights.
£90.21 - £1107.70
Specifications
For Use With (Application) | For use with Weighing Instruments of Medium Accuracy Class III and Ordinary Accuracy Class IIII |
---|
Product Code | Brand | Tolerance | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Tolerance | Price | Quantity & Availability | |||||
11790763
|
Zwiebel™
PBI23122 |
±5 mg |
£101.34
Each |
Estimated Shipment: 12-06-2024
Log in to see stock. |
|||||
11760763
|
Zwiebel™
PBI23119 |
±2 mg |
£93.17
Each |
Estimated Shipment: 23-05-2024
Log in to see stock. |
|||||
11730763
|
Zwiebel™
PBI23116 |
±1 mg |
£90.21
Each |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
11730773
|
Zwiebel™
PBI23126 |
±100 mg |
£359.34
Each |
Estimated Shipment: 12-06-2024
Log in to see stock. |
|||||
11700773
|
Zwiebel™
PBI23123 |
±10 mg |
£114.48
Each |
Estimated Shipment: 16-05-2024
Log in to see stock. |
|||||
11740763
|
Zwiebel™
PBI23117 |
±1.2 mg |
£90.52
Each |
Estimated Shipment: 12-06-2024
Log in to see stock. |
|||||
11750773
|
Zwiebel™
PBI23128 |
±500 mg |
£1107.70
Each |
Estimated Shipment: 12-06-2024
Log in to see stock. |
|||||
11740773
|
Zwiebel™
PBI23127 |
±250 mg |
£695.36
Each |
Estimated Shipment: 12-06-2024
Log in to see stock. |
|||||
11780763
|
Zwiebel™
PBI23121 |
±3 mg |
£97.10
Each |
Estimated Shipment: 12-06-2024
Log in to see stock. |
|||||
11750763
|
Zwiebel™
PBI23118 |
±1.6 mg |
£94.92
Each |
Estimated Shipment: 12-06-2024
Log in to see stock. |
|||||
Description
Specifications
For use with Weighing Instruments of Medium Accuracy Class III and Ordinary Accuracy Class IIII |