Learn More
Zwiebel™ Class M1 Box Weights with Certificate
Use with weighing instruments of medium accuracy class III. Zwiebel™ Class M1 Box Weights are ideal for the verification or calibration of class M₂ weights.
Brand: Zwiebel™ PBI23121
Code : A0
Additional Details : Weight : 0.80000kg
Description
Specifications
±3 mg | |
For use with Weighing Instruments of Medium Accuracy Class III and Ordinary Accuracy Class IIII |