missing translation for 'onlineSavingsMsg'
Learn More

Zwiebel™ Class M1 Box Weights with Certificate

Use with weighing instruments of medium accuracy class III. Zwiebel™ Class M1 Box Weights are ideal for the verification or calibration of class M₂ weights.

Brand:  Zwiebel™ PBI23128

Code : A0

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11750773

  • £1107.70 / Each
Estimated Shipment: 25-06-2024
to see stock.
Add to basket

Description

Description

  • Class M1 stainless steel calibration weight
  • Delivered with a calibration certificate guaranteeing traceability
  • In compliance with OIML R111
  • Supplied individually or as a set, in a plastic case or a wooden box
  • Specifications

    Specifications

    ±500 mg
    For use with Weighing Instruments of Medium Accuracy Class III and Ordinary Accuracy Class IIII
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    View accessory products
    Special Offers

    Special Offers