missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ CIRCLEGROW™ Capsules, Molecular Biology Certified
Designed for rapid growth of E. coli and high yields of plasmid DNA.
£198.22 - £995.34
Specifications
Content And Storage | 15°C to 30°C |
---|---|
Format | Capsule |
Product Type | Microbiology Media |
Formulation | Yeast extract, Tryptone broth, Vitamins, Biological Buffers have been added for proper pH adjustment |
For Use With (Application) | For growth of Escherichia coli and certified for plasmid and other bacterial cultures in liquid medium |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11313189
|
MP Biomedicals™
113000121 |
250 Capsules |
£198.22
Pack of 250 |
Estimated Shipment: 25-07-2024
Log in to see stock. |
|||||
11343189
|
MP Biomedicals™
113000131 |
1000 Capsules |
£995.34
Pack of 1000 |
Estimated Shipment: 26-07-2024
Log in to see stock. |
|||||
Description
- Encapsulated growth media for growth of E.coli
- Extends plasmid production past log phase
- Enhanced bacterial media
Specifications
15°C to 30°C | |
Microbiology Media | |
For growth of Escherichia coli and certified for plasmid and other bacterial cultures in liquid medium |
Capsule | |
Yeast extract, Tryptone broth, Vitamins, Biological Buffers have been added for proper pH adjustment |