missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ CIRCLEGROW™ Capsules, Molecular Biology Certified
Designed for rapid growth of E. coli and high yields of plasmid DNA.
Brand: MP Biomedicals™ 113000131
Code : 50
Additional Details : Weight : 0.73350kg
Description
- Encapsulated growth media for growth of E.coli
- Extends plasmid production past log phase/li>
- Enhanced bacterial media
Specifications
15°C to 30°C | |
Molecular biology | |
Yeast extract, Tryptone broth, Vitamins, Biological Buffers have been added for proper pH adjustment | |
1000 Capsules |
Capsule | |
Microbiology Media | |
For growth of Escherichia coli and certified for plasmid and other bacterial cultures in liquid medium |