missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ CIRCLEGROW™ Capsules, Molecular Biology Certified
Shop All MP Biomedicals Products
Click to view available options
Quantity:
1000 Capsules
250 Capsules
Unit Size:
Pack of 1000
Pack of 250
Description
- Encapsulated growth media for growth of E.coli
- Extends plasmid production past log phase/li>
- Enhanced bacterial media
Specifications
Specifications
| Content And Storage | 15°C to 30°C |
| Format | Capsule |
| Grade | Molecular biology grade |
| Product Type | Microbiology Media |
| Formulation | Yeast extract, Tryptone broth, Vitamins, Biological Buffers have been added for proper pH adjustment |
| For Use With (Application) | For growth of Escherichia coli and certified for plasmid and other bacterial cultures in liquid medium |
| Packaging Type | Container |
| Quantity | 250 Capsules |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction