missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech ChromoTek V5-Trap™ Agarose
V5-Trap™ Agarose is an affinity reagent for IP and purification of V5-tagged proteins.
£208.00 - £5160.00
Specifications
Content And Storage | Shipped at ambient temperature. Upon receipt store at 4°C. Stable for one year. |
---|---|
For Use With (Application) | IP, CoIP, ChIP, RIP |
Target | V5-tag sequence GKPIPNPLLGLDST at the N-, C-terminal or internal tagging. |
Label or Dye | Agarose |
Species | Alpaca |
Product Code | Brand | Sufficient For | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Sufficient For | Price | Quantity & Availability | |||||
17323533
|
Proteintech
V5TA-10 |
10 Reactions |
£208.00
10 reactions |
Estimated Shipment: 30-04-2024
Log in to see stock. |
|||||
17333533
|
Proteintech
V5TA-100 |
100 Reactions |
£1572.00
100 reactions |
Estimated Shipment: 21-06-2024
Log in to see stock. |
|||||
17343533
|
Proteintech
V5TA-20 |
20 Reactions |
£348.00
20 reactions |
Estimated Shipment: 21-06-2024
Log in to see stock. |
|||||
17353533
|
Proteintech
V5TA-200 |
200 Reactions |
£2828.00
200 reactions |
Estimated Shipment: 21-06-2024
Log in to see stock. |
|||||
17363533
|
Proteintech
V5TA-400 |
400 Reactions |
£5160.00
400 reactions |
Estimated Shipment: 21-06-2024
Log in to see stock. |
|||||
Description
- Consists of an anti-V5 Nanobody/ VHH coupled to agarose beads
Specifications
Shipped at ambient temperature. Upon receipt store at 4°C. Stable for one year. | |
V5-tag sequence GKPIPNPLLGLDST at the N-, C-terminal or internal tagging. | |
Alpaca |
IP, CoIP, ChIP, RIP | |
Agarose |