Learn More
Proteintech ChromoTek V5-Trap™ Agarose
V5-Trap™ Agarose is an affinity reagent for IP and purification of V5-tagged proteins.
Brand: Proteintech V5TA-100
Additional Details : Weight : 0.00120kg
Description
- Consists of an anti-V5 Nanobody/ VHH coupled to agarose beads
Specifications
Shipped at ambient temperature. Upon receipt store at 4°C. Stable for one year. | |
V5-tag sequence GKPIPNPLLGLDST at the N-, C-terminal or internal tagging. | |
Alpaca |
IP, CoIP, ChIP, RIP | |
Agarose | |
100 Reactions |