missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHO cells Rabbit anti-Hamster, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | CHO cells |
|---|---|
| Concentration | 1.33 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16865835
|
Proteintech
27803-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16855835
|
Proteintech
27803-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This antibody is raised by the total CHO cell lysate. It detects all the CHO proteins.
CHO cells (Chinese Hamster Ovary cells) are a laboratory-cultured cell line derived from cells of the ovaries of Chinese hamsters.Specifications
| CHO cells | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Hamster | |
| Chinese Hamster Ovary cells, CHO, CHO cells | |
| Primary | |
| -20°C |
| 1.33 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title