missing translation for 'onlineSavingsMsg'
Learn More

CHO cells Rabbit anti-Hamster, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 27803-1-AP-150UL

 View more versions of this product

Product Code. 16855835

  • £349.00 / 150µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This antibody is raised by the total CHO cell lysate. It detects all the CHO proteins.

CHO cells (Chinese Hamster Ovary cells) are a laboratory-cultured cell line derived from cells of the ovaries of Chinese hamsters.
Specifications

Specifications

CHO cells
Polyclonal
Unconjugated
Chinese Hamster Ovary cells, CHO, CHO cells
Antigen Affinity Chromatography
RUO
Hamster
Antibody
IgG
Western Blot
1.33 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Rabbit
150 μL
Primary
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.