missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHO cells Rabbit anti-Hamster, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27803-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody is raised by the total CHO cell lysate. It detects all the CHO proteins.
CHO cells (Chinese Hamster Ovary cells) are a laboratory-cultured cell line derived from cells of the ovaries of Chinese hamsters.Specifications
| CHO cells | |
| Polyclonal | |
| Unconjugated | |
| Chinese Hamster Ovary cells, CHO, CHO cells | |
| Antigen Affinity Chromatography | |
| RUO | |
| Hamster | |
| Antibody | |
| IgG |
| Western Blot | |
| 1.33 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Rabbit | |
| 20 μL | |
| Primary | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction