missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chemerin Antibody (13G2) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
£153.00 - £426.00
Specifications
| Antigen | Chemerin |
|---|---|
| Clone | 13G2 |
| Concentration | LYOPH |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18113094
|
Novus Biologicals
NBP2-21989-0.025MG |
0.025 mg |
£153.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18198651
|
Novus Biologicals
NBP2-21989 |
0.1 mg |
£426.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemerin Monoclonal specifically detects Chemerin in Mouse samples. It is validated for ELISA.Specifications
| Chemerin | |
| LYOPH | |
| Monoclonal | |
| Purified | |
| RUO | |
| Q9DD06 | |
| 5919 | |
| IgG2 | |
| Protein G purified |
| 13G2 | |
| ELISA | |
| Unconjugated | |
| Rat | |
| Mouse | |
| chemerin, RAR-responsive protein TIG2, retinoic acid receptor responder (tazarotene induced) 2, retinoic acid receptor responder protein 2, Tazarotene-induced gene 2 protein, TIG2HP10433 | |
| RARRES2 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title