missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chemerin Antibody (13G2) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-21989-0.025MG
This item is not returnable.
View return policy
Description
Chemerin Monoclonal specifically detects Chemerin in Mouse samples. It is validated for ELISA.
Specifications
| Chemerin | |
| Monoclonal | |
| LYOPH | |
| ELISA 1:2000 | |
| Q9DD06 | |
| RARRES2 | |
| This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with mouse recombinant protein Chemerin. Rat monoclonal anti mouse Chemerin Q9DD06. | |
| 0.025 mg | |
| Primary | |
| Reconstitute with 0.5 ml sterilized PBS to a final concentration of 0.2 mg/ml. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2 |
| ELISA | |
| 13G2 | |
| Unconjugated | |
| A 0.2 um filtered solution in PBS with No Preservative | |
| chemerin, RAR-responsive protein TIG2, retinoic acid receptor responder (tazarotene induced) 2, retinoic acid receptor responder protein 2, Tazarotene-induced gene 2 protein, TIG2HP10433 | |
| Rat | |
| Protein G purified | |
| RUO | |
| 5919 | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction