missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chemerin Antibody (13G2) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-21989
This item is not returnable.
View return policy
Description
Chemerin Monoclonal specifically detects Chemerin in Mouse samples. It is validated for ELISA.
Specifications
| Chemerin | |
| Monoclonal | |
| LYOPH | |
| ELISA 1:1000-1:2000 | |
| Q9DD06 | |
| RARRES2 | |
| Mouse recombinant Chemerin | |
| 0.1 mg | |
| Cardiovascular Biology, metabolism | |
| 5919 | |
| Mouse | |
| Purified |
| ELISA | |
| 13G2 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS. 0.025 mg size is provided in liquid form, PBS with No Preservative | |
| chemerin, RAR-responsive protein TIG2, retinoic acid receptor responder (tazarotene induced) 2, retinoic acid receptor responder protein 2, Tazarotene-induced gene 2 protein, TIG2HP10433 | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| Reconstitute with sterilized PBS to a final concentration of 0.2 mg/ml. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction