missing translation for 'onlineSavingsMsg'
Learn More

Cefsulodin sodium salt hydrate, 97%, Thermo Scientific™

Cefsulodin sodium (sulcephalosporin, CAS # 52152-93-9) is the salt of the semi-synthetic cephalosporin antibiotic.

£74.30 - £235.00

Chemical Identifiers

CAS 1426397-23-0
Molecular Formula C22H21N4NaO8S2+
Molecular Weight (g/mol) 556.54
MDL Number MFCD07793338
InChI Key GTZPOHRNWUTXNB-DWBVFMGKSA-O
PubChem CID 124203950
IUPAC Name (6R,7R)-3-[(4-carbamoylpyridin-1-ium-1-yl)methyl]-8-oxo-7-[[(2R)-2-phenyl-2-sulfoacetyl]amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;sodium
SMILES C1C(=C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=CC=C3)S(=O)(=O)O)C(=O)O)C[N+]4=CC=C(C=C4)C(=O)N.[Na]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15427369
SDS View Documents
Thermo Scientific Alfa Aesar
J66120.MD
250mg
£74.30
250mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15497359
SDS View Documents
Thermo Scientific Alfa Aesar
J66120.03
1g
£235.00
1g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

General Description

• Cefsulodin sodium is a third-generation cephalosporin antibiotic with a narrow range of effectiveness
• Cefsulodin competitively inhibits penicillin binding protein, preventing cell wall synthesis and promoting lysis

Application

• Cefsulodin is mainly effective against Yersinia microorganisms, Pseudomonas aeruginosa, and Staphylococcus aureus
• Cefsulodin is useful for drug resistance studies involving susceptible microbes and for studies related to drug mechanisms
• It is appropriate for selection experiments and other cell culture applications

Specifications

Chemical Identifiers

1426397-23-0
556.54
GTZPOHRNWUTXNB-DWBVFMGKSA-O
(6R,7R)-3-[(4-carbamoylpyridin-1-ium-1-yl)methyl]-8-oxo-7-[[(2R)-2-phenyl-2-sulfoacetyl]amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;sodium
C22H21N4NaO8S2+
MFCD07793338
124203950
C1C(=C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=CC=C3)S(=O)(=O)O)C(=O)O)C[N+]4=CC=C(C=C4)C(=O)N.[Na]

Specifications

1426397-23-0
C22H21N4NaO8S2+
14,1945
GTZPOHRNWUTXNB-DWBVFMGKSA-O
(6R,7R)-3-[(4-carbamoylpyridin-1-ium-1-yl)methyl]-8-oxo-7-[[(2R)-2-phenyl-2-sulfoacetyl]amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;sodium
124203950
97%
White to Yellow
MFCD07793338
Freely soluble in water
C1C(=C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=CC=C3)S(=O)(=O)O)C(=O)O)C[N+]4=CC=C(C=C4)C(=O)N.[Na]
556.54
554.53 (Anhydrous)
Cefsulodin sodium salt hydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H334-H335-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.

GHS P Statement
P285-P261-P280-P305+P351+P338-P405-P501a
In case of inadequate ventilation wear respiratory protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

missing translation for 'rtecsNumber' : UU1785000

missing translation for 'tsca' : TSCA

missing translation for 'storageNote' : -30° to -10°C

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.