Learn More
Cefsulodin sodium salt hydrate, 97%, Thermo Scientific™
Cefsulodin sodium (sulcephalosporin, CAS # 52152-93-9) is the salt of the semi-synthetic cephalosporin antibiotic.
Brand: Thermo Scientific Alfa Aesar J66120.MD
| Quantity | 250mg |
|---|
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
General Description
• Cefsulodin sodium is a third-generation cephalosporin antibiotic with a narrow range of effectiveness
• Cefsulodin competitively inhibits penicillin binding protein, preventing cell wall synthesis and promoting lysis
Application
• Cefsulodin is mainly effective against Yersinia microorganisms, Pseudomonas aeruginosa, and Staphylococcus aureus
• Cefsulodin is useful for drug resistance studies involving susceptible microbes and for studies related to drug mechanisms
• It is appropriate for selection experiments and other cell culture applications
Chemical Identifiers
| 1426397-23-0 | |
| 556.54 | |
| GTZPOHRNWUTXNB-DWBVFMGKSA-O | |
| (6R,7R)-3-[(4-carbamoylpyridin-1-ium-1-yl)methyl]-8-oxo-7-[[(2R)-2-phenyl-2-sulfoacetyl]amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;sodium |
| C22H21N4NaO8S2+ | |
| MFCD07793338 | |
| 124203950 | |
| C1C(=C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=CC=C3)S(=O)(=O)O)C(=O)O)C[N+]4=CC=C(C=C4)C(=O)N.[Na] |
Specifications
| 1426397-23-0 | |
| 250mg | |
| MFCD07793338 | |
| Freely soluble in water | |
| C1C(=C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=CC=C3)S(=O)(=O)O)C(=O)O)C[N+]4=CC=C(C=C4)C(=O)N.[Na] | |
| 556.54 | |
| 554.53 (Anhydrous) | |
| Cefsulodin sodium salt hydrate |
| White to Yellow | |
| C22H21N4NaO8S2+ | |
| 14,1945 | |
| GTZPOHRNWUTXNB-DWBVFMGKSA-O | |
| (6R,7R)-3-[(4-carbamoylpyridin-1-ium-1-yl)methyl]-8-oxo-7-[[(2R)-2-phenyl-2-sulfoacetyl]amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;sodium | |
| 124203950 | |
| 97% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.