CAS RN 1426397-23-0
CAS RN 1426397-23-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Cefsulodin sodium salt hydrate, 97%, Thermo Scientific™
CAS: 1426397-23-0 Molecular Formula: C22H21N4NaO8S2+ Molecular Weight (g/mol): 556.54 MDL Number: MFCD07793338 InChI Key: GTZPOHRNWUTXNB-DWBVFMGKSA-O PubChem CID: 124203950 IUPAC Name: (6R,7R)-3-[(4-carbamoylpyridin-1-ium-1-yl)methyl]-8-oxo-7-[[(2R)-2-phenyl-2-sulfoacetyl]amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;sodium SMILES: C1C(=C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=CC=C3)S(=O)(=O)O)C(=O)O)C[N+]4=CC=C(C=C4)C(=O)N.[Na]