missing translation for 'onlineSavingsMsg'
Learn More

Cdk4 Ab-1 Mouse Monoclonal Antibody, Epredia™

Ensure accurate and reproducible results in western blotting, immunofluorescence, immunoprecipitation and kinase assay applications.

£119.34 - £584.00

Specifications

Antigen Cdk4 Ab-1
Clone DCS-35
Applications Immunofluorescence, Immunoprecipitation, Western Blot
Classification Monoclonal
Form Concentrated
View More Specs

Products 7
Product Code Brand Quantity Conjugate Concentration Price Quantity & Availability  
Product Code Brand Quantity Conjugate Concentration Price Quantity & Availability  
12670877
View Documents
Epredia
MS-299-B0
100 μL Biotin 200μg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12660877
View Documents
Epredia
MS-299-B1
500 μL Biotin 200μg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12690877
View Documents
Epredia
MS-299-P0
100 μL Unconjugated 200μg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12600887
View Documents
Epredia
MS-299-P1
500 μL Unconjugated 200μg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12680877
View Documents
Epredia
MS-299-P
1 mL Unconjugated 200μg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12610887
View Documents
Epredia
MS-299-P1ABX
100 μL Unconjugated 1mg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12620887
View Documents
Epredia
MS-299-PABX
200 μL Unconjugated 1mg/mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

A family of proteins designated as cdks are critical regulators of cell cycle progression. The prototype member of this family, p34cdc2, and a related protein cdk2, function late in the cycle while cdk4 and cdk6 are critically involved in G1 to S progression.

Host Species: Mouse

Clone: DCS-35

Isotype: IgG1/κ

Species Reactivity: Human, Mouse, Rat and Pig. Others not known.

Epitope: aa 1-20

Immunogen: Purified recombinant Cdk4 protein

Molecular Weight: 34 kDa

Positive Control: NIH 3T3, MAD109, LS174T, or MCF-7 cells

Cellular Localization: Nuclear and cytoplasmic

Recommended for:

  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native only)
  • Kinase Assay

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Cdk4 Ab-1
Immunofluorescence, Immunoprecipitation, Western Blot
Concentrated
RUO
Human, Mouse, Pig, Rat
IgG2a κ
DCS-35
Monoclonal
Mouse
Cell Cycle
Purified recombinant cdk4 protein
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.