missing translation for 'onlineSavingsMsg'
Learn More

Cdk4 Ab-1 Mouse Monoclonal Antibody, Epredia™

Ensure accurate and reproducible results in western blotting, immunofluorescence, immunoprecipitation and kinase assay applications.

Brand:  Epredia MS-299-PABX

 View more versions of this product

Product Code. 12620887

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

A family of proteins designated as cdks are critical regulators of cell cycle progression. The prototype member of this family, p34cdc2, and a related protein cdk2, function late in the cycle while cdk4 and cdk6 are critically involved in G1 to S progression.

Host Species: Mouse

Clone: DCS-35

Isotype: IgG1/κ

Species Reactivity: Human, Mouse, Rat and Pig. Others not known.

Epitope: aa 1-20

Immunogen: Purified recombinant Cdk4 protein

Molecular Weight: 34 kDa

Positive Control: NIH 3T3, MAD109, LS174T, or MCF-7 cells

Cellular Localization: Nuclear and cytoplasmic

Recommended for:

  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native only)
  • Kinase Assay

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cdk4 Ab-1
Monoclonal
1mg/mL
Mouse
200 μL
Cell Cycle
Human, Mouse, Pig, Rat
IgG2a κ
Immunofluorescence, Immunoprecipitation, Western Blot
DCS-35
Unconjugated
Purified recombinant cdk4 protein
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.