Learn More
Cdk4 Ab-1 Mouse Monoclonal Antibody, Epredia™
Ensure accurate and reproducible results in western blotting, immunofluorescence, immunoprecipitation and kinase assay applications.
Brand: Epredia MS-299-P
Description
A family of proteins designated as cdks are critical regulators of cell cycle progression. The prototype member of this family, p34cdc2, and a related protein cdk2, function late in the cycle while cdk4 and cdk6 are critically involved in G1 to S progression.Host Species: Mouse
Clone: DCS-35
Isotype: IgG1/κ
Species Reactivity: Human, Mouse, Rat and Pig. Others not known.
Epitope: aa 1-20
Immunogen: Purified recombinant Cdk4 protein
Molecular Weight: 34 kDa
Positive Control: NIH 3T3, MAD109, LS174T, or MCF-7 cells
Cellular Localization: Nuclear and cytoplasmic
Recommended for:
- Western Blotting
- Immunofluorescence
- Immunoprecipitation (Native only)
- Kinase Assay
Note:
USA: RUO; Int'l: RUO
Specifications
| Cdk4 Ab-1 | |
| Monoclonal | |
| 200μg/mL | |
| Mouse | |
| 1 mL | |
| Cell Cycle | |
| Human, Mouse, Pig, Rat | |
| IgG2a κ |
| Immunofluorescence, Immunoprecipitation, Western Blot | |
| DCS-35 | |
| Unconjugated | |
| Purified recombinant cdk4 protein | |
| RUO | |
| Primary | |
| Concentrated |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.