missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD83 Mouse, anti-Human, APC, Clone: HB15e, STEMCELL Technologies
Mouse Monoclonal Antibody
£283.50 - £608.85
Specifications
| Antigen | CD83 |
|---|---|
| Clone | HB15e |
| Applications | Immunoblot, Flow Cytometry, Immunohistochemistry |
| Classification | Monoclonal |
| Conjugate | APC |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17119061
|
Stemcell Technologies
60107AZ.1 |
25 Tests |
£283.50
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17109061
|
Stemcell Technologies
60107AZ |
100 Tests |
£608.85
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- The HB15e antibody reacts with CD83, an ∼45 kDa type I transmembrane glycoprotein and member of the Siglec protein family, containing an extracellular V-type Ig-like domain and a C-terminal cytoplasmic tail
- A soluble form of CD83 has also been identified
- CD83 is expressed predominantly on circulating and tissue dendritic cells, including Langerhan cells and interdigitating reticulum cells
- It is expressed weakly on activated lymphocytes and may also be found on neutrophils, monocytes and macrophages
- Expression of CD83 is typically transient, except on dendritic cells, and correlates with upregulation of HLA class II antigen
- Its function, though ill-defined, is essential for the development and maintenance of the immune system and evidence suggests it may be involved in cell adhesion and signaling, antigen presentation, and lymphocyte activation
- The epitope on CD83 bound by the HB15e antibody reportedly overlaps with that of antibody 1E11
Specifications
| CD83 | |
| Immunoblot, Flow Cytometry, Immunohistochemistry | |
| APC | |
| Baboon, Cynomolgus, Rhesus Monkey, Chimpanzee, Human | |
| Primary |
| HB15e | |
| Monoclonal | |
| Mouse | |
| IgG1 κ |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title