Learn More
CD83 Mouse, anti-Human, APC, Clone: HB15e, STEMCELL Technologies
Mouse Monoclonal Antibody
Brand: Stemcell Technologies 60107AZ.1
This item is not returnable.
View return policy
Description
- The HB15e antibody reacts with CD83, an ∼45 kDa type I transmembrane glycoprotein and member of the Siglec protein family, containing an extracellular V-type Ig-like domain and a C-terminal cytoplasmic tail
- A soluble form of CD83 has also been identified
- CD83 is expressed predominantly on circulating and tissue dendritic cells, including Langerhan cells and interdigitating reticulum cells
- It is expressed weakly on activated lymphocytes and may also be found on neutrophils, monocytes and macrophages
- Expression of CD83 is typically transient, except on dendritic cells, and correlates with upregulation of HLA class II antigen
- Its function, though ill-defined, is essential for the development and maintenance of the immune system and evidence suggests it may be involved in cell adhesion and signaling, antigen presentation, and lymphocyte activation
- The epitope on CD83 bound by the HB15e antibody reportedly overlaps with that of antibody 1E11
Specifications
| CD83 | |
| Monoclonal | |
| APC | |
| 25 Tests | |
| Baboon, Cynomolgus, Rhesus Monkey, Chimpanzee, Human | |
| Dendritic Cells |
| Immunoblot, Flow Cytometry, Immunohistochemistry | |
| HB15e | |
| Mouse | |
| Primary | |
| IgG1 κ |