missing translation for 'onlineSavingsMsg'
Learn More

IBA Lifesciences CD8 Isolation Kit for FABian™

IBA Lifesciences™ CD8 Isolation Kit for FABian™ is for reversible cell isolation delivering label-free target cells.

Brand:  IBA Lifesciences 6-6003-010

Additional Details : Weight : 1.70000kg

 View more versions of this product

Product Code. 15879774

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

  • The kit is used for the selection of CD8+ T cells from whole blood or buffy coat in conjunction with the FABian™ system.
  • It includes CD8 Fab-Strep (human), the Cell Isolation Column, Cell Isolation Buffer, Biotin for Cell Elution and tubes and a luer lock adapter with the syringe.
  • Each kit is sufficient for one FABian™ run to isolate cells from 6.25mL buffy coat.
  • Alternatively 6.25mL whole blood may be used.
Specifications

Specifications

Isolation of label-free cells
2°C to 8°C
9 mL Whole Blood
Non-magnetic Cell Isolation Kit
Human Whole Blood
10 Kits, Human
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers