missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IBA Lifesciences CD8 Isolation Kit for FABian™
IBA Lifesciences™ CD8 Isolation Kit for FABian™ is for reversible cell isolation delivering label-free target cells.
£84.14 - £664.86
Specifications
Description | Isolation of label-free cells |
---|---|
Product Type | Non-magnetic Cell Isolation Kit |
Content And Storage | 2°C to 8°C |
Donor Source | Human Whole Blood |
Sufficient For | 9 mL Whole Blood |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15879774
|
IBA Lifesciences
6-6003-010 |
10 Kits, Human |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15869774
|
IBA Lifesciences
6-6003-001 |
1 Kit, Human |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- The kit is used for the selection of CD8+ T cells from whole blood or buffy coat in conjunction with the FABian™ system.
- It includes CD8 Fab-Strep (human), the Cell Isolation Column, Cell Isolation Buffer, Biotin for Cell Elution and tubes and a luer lock adapter with the syringe.
- Each kit is sufficient for one FABian™ run to isolate cells from 6.25mL buffy coat.
- Alternatively 6.25mL whole blood may be used.
Specifications
Isolation of label-free cells | |
2°C to 8°C | |
9 mL Whole Blood |
Non-magnetic Cell Isolation Kit | |
Human Whole Blood |